siRNA Details
Gene Symbol: USP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020935.2 | NP_065986.2 | 17 | 2114 | ||
XM_005246720.2 | XP_005246777.1 | 17 | 2186 | |||
XM_005246721.3 | XP_005246778.1 | 17 | 2078 | |||
XM_005246722.3 | XP_005246779.1 | 17 | 2118 | |||
XM_005246724.1 | XP_005246781.1 | 17 | 2114 | |||
XM_011511538.2 | XP_011509840.1 | 16 | 2059 | |||
XM_011511541.2 | XP_011509843.1 | 17 | 2114 | |||
XM_017004577.1 | XP_016860066.1 | 16 | 1974 | |||
GenBank mRNA | AB046814.1 | - | 13 | 1559 | ||
AK095577.1 | - | 17 | 2072 | |||
AK128583.1 | - | 17 | 2118 | |||
AL832645.2 | - | 17 | 2113 | |||
BC112901.1 | - | 16 | 1914 | 6016503 | ||
BC133007.1 | - | 17 | 1999 | 40147029 | ||
BC133009.1 | - | 17 | 2044 | 40147031 | ||
BC144249.1 | - | 17 | 1999 | 9052769 | ||
BX538024.1 | - | 17 | 2079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
