siRNA Details
Gene Symbol: PRDM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021620.3 | NP_067633.2 | 4 | 3029 | ||
GenBank mRNA | AY004253.1 | - | 4 | 3059 | ||
AY130010.1 | - | 1 | 1272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
