siRNA Details
Gene Symbol: SMYD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167740.1 | NP_001161212.1 | 4 | 479 | ||
NM_022743.2 | NP_073580.1 | 4 | 307 | |||
XM_011544253.1 | XP_011542555.1 | 4 | 455 | |||
GenBank mRNA | AB057595.1 | - | 4 | 449 | ||
AK024733.1 | - | 4 | 304 | |||
AK289605.1 | - | 4 | 455 | |||
BC031010.1 | - | 4 | 394 | 4334047 | ||
BC107725.1 | - | 4 | 453 | |||
BI916760.1 | - | 4 | 371 | |||
BP231249.1 | - | 4 | 478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
