siRNA Details
Gene Symbol: ARFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025593.2 | NP_001020764.1 | 3, 4 | 509 | ||
NM_001287433.1 | NP_001274362.1 | 4, 5 | 491 | |||
NM_014447.3 | NP_055262.1 | 3, 4 | 374 | |||
XM_011531871.2 | XP_011530173.1 | 2, 3 | 3338 | |||
GenBank mRNA | AK096509.1 | - | 3, 4 | 458 | ||
AK290472.1 | - | 3, 4 | 345 | |||
BC105114.1 | - | 3, 4 | 461 | 8144117 | ||
BC105116.1 | - | 3, 4 | 461 | 8144119 | ||
BC143675.1 | - | 3, 4 | 461 | 9052189 | ||
CN305403.1 | - | 3, 4 | 457 | |||
DA452239.1 | - | 4, 5 | 483 | |||
U52521.1 | - | 3, 4 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
