siRNA Details
Gene Symbol: WHSC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017778.2 | NP_060248.2 | 6 | 1742 | ||
NM_023034.1 | NP_075447.1 | 6 | 1742 | |||
GenBank mRNA | AF255649.1 | - | 7 | 1344 | ||
AF332468.1 | - | 6 | 1742 | |||
AF332469.1 | - | 6 | 1742 | |||
AJ295990.1 | - | 6 | 1537 | |||
AJ295991.1 | - | 6 | 1537 | |||
AJ295992.1 | - | 6 | 1537 | |||
AK000360.1 | - | 1 | 41 | |||
AK022560.1 | - | 6 | 1419 | |||
BC012059.1 | - | 6 | 1710 | |||
BC062631.1 | - | 6 | 1681 | |||
BC073858.1 | - | 5 | 1268 | |||
BC101717.1 | - | 6 | 1316 | 8069223 | ||
BC113469.1 | - | 6 | 1316 | 8322521 | ||
BC115006.1 | - | 6 | 1337 | 7961962 | ||
BC143510.1 | - | 6 | 1316 | 9052018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
