siRNA Details
Gene Symbol: WHSC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017778.2 | NP_060248.2 | 6, 7 | 2107 | ||
NM_023034.1 | NP_075447.1 | 6, 7 | 2107 | |||
GenBank mRNA | AF255649.1 | - | 7, 8 | 1709 | ||
AF332468.1 | - | 6, 7 | 2107 | |||
AF332469.1 | - | 6, 7 | 2107 | |||
AJ295990.1 | - | 6, 7 | 1902 | |||
AJ295991.1 | - | 6, 7 | 1902 | |||
AJ295992.1 | - | 6, 7 | 1902 | |||
AK000360.1 | - | 1, 2 | 406 | |||
AK022560.1 | - | 6, 7 | 1784 | |||
BC012059.1 | - | 6, 7 | 2075 | |||
BC062631.1 | - | 6, 7 | 2046 | |||
BC073858.1 | - | 5, 6 | 1633 | |||
BC101717.1 | - | 6, 7 | 1681 | 8069223 | ||
BC113469.1 | - | 6, 7 | 1681 | 8322521 | ||
BC115006.1 | - | 6, 7 | 1702 | 7961962 | ||
BC143510.1 | - | 6, 7 | 1681 | 9052018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
