siRNA Details
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193424.1 | NP_001180353.1 | 6 | 2706 | ||
NM_001193425.1 | NP_001180354.1 | 6 | 2664 | |||
NM_001193426.1 | NP_001180355.1 | 6 | 2166 | |||
NM_001193427.1 | NP_001180356.1 | 6 | 2124 | |||
NM_024670.3 | NP_078946.1 | 5 | 2651 | |||
NR_034181.1 | 5 | 2560 | ||||
XM_011519662.2 | XP_011517964.1 | 6 | 2695 | |||
XM_017016637.1 | XP_016872126.1 | 5 | 2601 | |||
GenBank mRNA | AK027067.1 | - | 5 | 2457 | ||
BC007754.2 | - | 5 | 2651 | 4109807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
