siRNA Details
Gene Symbol: SETD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160305.2 | NP_001153777.1 | 8 | 1605 | ||
NM_024860.3 | NP_079136.2 | 9 | 1533 | |||
NR_134583.1 | 9 | 1564 | ||||
GenBank mRNA | AK024801.1 | - | 9 | 1489 | ||
AK127136.1 | - | 5 | 3010 | |||
BC110433.1 | - | 9 | 1548 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
