siRNA Details
Gene Symbol: USP36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321291.1 | NP_001308220.1 | 21 | 3842 | ||
XM_005257542.2 | XP_005257599.1 | 21 | 3866 | |||
XM_005257543.2 | XP_005257600.1 | 21 | 3889 | |||
XM_011525065.1 | XP_011523367.1 | 21 | 3903 | |||
XM_011525066.2 | XP_011523368.1 | 21 | 3742 | |||
XM_011525068.1 | XP_011523370.1 | 21 | 3806 | |||
XM_011525069.2 | XP_011523371.1 | 20 | 3168 | |||
XM_011525070.1 | XP_011523372.1 | 22 | 3836 | |||
XM_017024901.1 | XP_016880390.1 | 19 | 3427 | |||
GenBank mRNA | AB040886.1 | - | 20 | 4273 | ||
AK022840.1 | - | 16 | 3806 | |||
AK023077.1 | - | 3 | 401 | |||
AL833835.1 | - | 7 | 2580 | |||
BC013890.2 | - | 6 | 763 | |||
BC071582.1 | - | 21 | 3842 | 30345255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
