siRNA Details
Gene Symbol: ITCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257137.2 | NP_001244066.1 | 10 | 1167 | ||
NM_001257138.2 | NP_001244067.1 | 8 | 902 | |||
NM_001324197.1 | NP_001311126.1 | 10 | 1188 | |||
NM_001324198.1 | NP_001311127.1 | 9 | 1065 | |||
NM_031483.6 | NP_113671.3 | 9 | 1044 | |||
XM_005260570.1 | XP_005260627.1 | 8 | 960 | |||
XM_011529079.1 | XP_011527381.1 | 8 | 980 | |||
XM_017028089.1 | XP_016883578.1 | 9 | 1000 | |||
XM_017028090.1 | XP_016883579.1 | 10 | 1120 | |||
XM_017028091.1 | XP_016883580.1 | 8 | 915 | |||
XM_017028092.1 | XP_016883581.1 | 10 | 1120 | |||
GenBank mRNA | AB056663.2 | - | 9 | 954 | ||
AB209747.1 | - | 1 | 61 | |||
AF038564.1 | - | 4 | 462 | |||
AF095745.1 | - | 7 | 837 | |||
AK297809.1 | - | 9 | 862 | |||
AK304090.1 | - | 8 | 861 | |||
AK315212.1 | - | 9 | 1003 | |||
BC011571.1 | - | 10 | 1090 | 4560366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
