siRNA Details
Gene Symbol: SETDB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160308.2 | NP_001153780.1 | 9 | 2123 | ||
NM_001320699.1 | NP_001307628.1 | 9 | 1802 | |||
NM_031915.2 | NP_114121.2 | 10 | 2159 | |||
GenBank mRNA | AB209349.1 | - | 2 | 4365 | ||
AF334407.1 | - | 10 | 2078 | |||
AK307593.1 | - | 8 | 1472 | |||
AK308220.1 | - | 9 | 2042 | |||
AL831937.1 | - | 3 | 260 | |||
BC028202.1 | - | 8 | 1596 | |||
BC047434.2 | - | 8 | 1559 | 5266911 |
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 9 | 2123 | ||
NR_135324.1 | 10 | 2289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
