siRNA Details
Gene Symbol: SETD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032233.2 | NP_115609.2 | 6 | 801 | ||
NM_199123.1 | NP_954574.1 | 6 | 801 | |||
XM_005268127.3 | XP_005268184.1 | 5 | 654 | |||
XM_011537231.2 | XP_011535533.2 | 6 | 753 | |||
XM_011537232.2 | XP_011535534.1 | 6 | 1931 | |||
XM_011537233.2 | XP_011535535.1 | 6 | 753 | |||
XM_011537234.1 | XP_011535536.1 | 5 | 521 | |||
XM_011537235.1 | XP_011535537.1 | 6 | 753 | |||
XM_017021699.1 | XP_016877188.1 | 5 | 649 | |||
XM_017021700.1 | XP_016877189.1 | 6 | 837 | |||
XM_017021701.1 | XP_016877190.1 | 6 | 803 | |||
XM_017021702.1 | XP_016877191.1 | 5 | 622 | |||
GenBank mRNA | AK026680.1 | - | 5 | 555 | ||
AK302882.1 | - | 6 | 2144 | |||
BC009054.1 | - | 3 | 427 | |||
BC066931.1 | - | 6 | 702 | |||
BC120967.1 | - | 6 | 656 | |||
BC120968.2 | - | 6 | 656 | |||
BC127624.1 | - | 6 | 656 | |||
BC127625.1 | - | 6 | 656 | |||
BC142995.1 | - | 6 | 813 | 8860450 | ||
BC148251.1 | - | 5 | 652 | |||
BX161441.1 | - | 6 | 700 | |||
BX161471.1 | - | 6 | 791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
