siRNA Details
Gene Symbol: USP48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032730.1 | NP_001027902.1 | 7 | 1036 | ||
NM_032236.5 | NP_115612.4 | 7 | 1036 | |||
XM_005246009.2 | XP_005246066.1 | 7 | 1040 | |||
XM_006710951.2 | XP_006711014.2 | 7 | 1040 | |||
XM_006710955.3 | XP_006711018.1 | 7 | 1016 | |||
XM_011542262.2 | XP_011540564.1 | 7 | 933 | |||
XM_011542263.2 | XP_011540565.1 | 7 | 933 | |||
XM_011542264.1 | XP_011540566.1 | 6 | 630 | |||
XM_011542265.2 | XP_011540567.1 | 8 | 1596 | |||
XM_011542267.2 | XP_011540569.1 | 7 | 933 | |||
XM_017002495.1 | XP_016857984.1 | 7 | 1040 | |||
XM_017002496.1 | XP_016857985.1 | 7 | 1040 | |||
GenBank mRNA | AF502942.1 | - | 7 | 938 | ||
AK074247.1 | - | 7 | 933 | |||
AK226150.1 | - | 7 | 1032 | |||
AK315005.1 | - | 7 | 816 | |||
BC067261.1 | - | 7 | 1004 | |||
BC103724.1 | - | 7 | 978 | |||
BC104896.1 | - | 7 | 832 | 8143899 | ||
BC143343.1 | - | 7 | 832 | 9051848 | ||
BC143349.1 | - | 7 | 832 | 9051854 | ||
BC143354.1 | - | 7 | 832 | 9051859 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
