siRNA Details
Gene Symbol: USP48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032730.1 | NP_001027902.1 | 11 | 1590 | ||
NM_032236.5 | NP_115612.4 | 11 | 1590 | |||
XM_005246009.2 | XP_005246066.1 | 11 | 1594 | |||
XM_006710951.2 | XP_006711014.2 | 11 | 1591 | |||
XM_006710955.3 | XP_006711018.1 | 11 | 1570 | |||
XM_011542262.2 | XP_011540564.1 | 11 | 1484 | |||
XM_011542263.2 | XP_011540565.1 | 11 | 1487 | |||
XM_011542264.1 | XP_011540566.1 | 10 | 1184 | |||
XM_011542265.2 | XP_011540567.1 | 12 | 2150 | |||
XM_011542267.2 | XP_011540569.1 | 11 | 1503 | |||
XM_011542268.2 | XP_011540570.1 | 3 | 203 | |||
XM_017002495.1 | XP_016857984.1 | 11 | 1591 | |||
XM_017002496.1 | XP_016857985.1 | 11 | 1594 | |||
XM_017002497.1 | XP_016857986.1 | 3 | 248 | |||
GenBank mRNA | AF502942.1 | - | 11 | 1492 | ||
AK074247.1 | - | 11 | 1484 | |||
AK226150.1 | - | 11 | 1586 | |||
AK315005.1 | - | 11 | 1370 | |||
BC007326.2 | - | 2 | 158 | |||
BC067261.1 | - | 11 | 1558 | |||
BC103724.1 | - | 11 | 1532 | |||
BC104896.1 | - | 11 | 1386 | 8143899 | ||
BC143343.1 | - | 11 | 1386 | 9051848 | ||
BC143349.1 | - | 11 | 1386 | 9051854 | ||
BC143354.1 | - | 11 | 1383 | 9051859 | ||
BM462540.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
