siRNA Details
Gene Symbol: USP38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290325.1 | NP_001277254.1 | 3 | 1437 | ||
NM_001290326.1 | NP_001277255.1 | 3 | 1437 | |||
NM_032557.6 | NP_115946.2 | 3 | 1437 | |||
XM_011532360.2 | XP_011530662.1 | 3 | 925 | |||
GenBank mRNA | AB067478.1 | - | 2 | 118 | ||
AK093754.1 | - | 3 | 660 | |||
AK126943.1 | - | 4 | 1265 | |||
AL833976.1 | - | 3 | 1409 | |||
BC039115.1 | - | 3 | 1437 | 5258804 | ||
BC068975.1 | - | 3 | 1384 | 6067501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
