siRNA Details
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 7, 8 | 971 | ||
NM_032663.4 | NP_116052.2 | 4, 5 | 597 | |||
XM_005253962.3 | XP_005254019.1 | 4, 5 | 583 | |||
XM_005253965.4 | XP_005254022.1 | 3, 4 | 408 | |||
XM_006719653.3 | XP_006719716.1 | 5, 6 | 670 | |||
XM_011538894.2 | XP_011537196.1 | 2, 3 | 304 | |||
XM_017020048.1 | XP_016875537.1 | 4, 5 | 592 | |||
XM_017020049.1 | XP_016875538.1 | 4, 5 | 583 | |||
XM_017020050.1 | XP_016875539.1 | 4, 5 | 692 | |||
XM_017020051.1 | XP_016875540.1 | 5, 6 | 548 | |||
XM_017020052.1 | XP_016875541.1 | 4, 5 | 594 | |||
XM_017020053.1 | XP_016875542.1 | 3, 4 | 324 | |||
XM_017020054.1 | XP_016875543.1 | 2, 3 | 202 | |||
GenBank mRNA | AJ586136.1 | - | 4, 5 | 488 | ||
AK027820.1 | - | 4, 5 | 562 | |||
AK097830.1 | - | 7, 8 | 971 | |||
BC004868.2 | - | 4, 5 | 587 | 3833242 | ||
BC022094.2 | - | 2, 3 | 139 | |||
HY127211.1 | - | 2, 3 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
