siRNA Details
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 12 | 1304 | ||
NM_032663.4 | NP_116052.2 | 9 | 930 | |||
XM_005253962.3 | XP_005254019.1 | 9 | 913 | |||
XM_005253965.4 | XP_005254022.1 | 8 | 741 | |||
XM_006719653.3 | XP_006719716.1 | 10 | 1003 | |||
XM_011538894.2 | XP_011537196.1 | 7 | 634 | |||
XM_017020048.1 | XP_016875537.1 | 9 | 925 | |||
XM_017020049.1 | XP_016875538.1 | 9 | 913 | |||
XM_017020050.1 | XP_016875539.1 | 9 | 1025 | |||
XM_017020051.1 | XP_016875540.1 | 10 | 881 | |||
XM_017020052.1 | XP_016875541.1 | 9 | 927 | |||
XM_017020053.1 | XP_016875542.1 | 8 | 657 | |||
XM_017020054.1 | XP_016875543.1 | 7 | 532 | |||
GenBank mRNA | AJ586136.1 | - | 9 | 821 | ||
AK027820.1 | - | 9 | 895 | |||
AK097830.1 | - | 12 | 1304 | |||
BC004868.2 | - | 9 | 920 | 3833242 | ||
BC022094.2 | - | 7 | 472 | |||
CN367077.1 | - | 3 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
