siRNA Details
Gene Symbol: USP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301175.1 | NP_001288104.1 | 14 | 1592 | ||
NM_032663.4 | NP_116052.2 | 11 | 1218 | |||
XM_005253962.3 | XP_005254019.1 | 11 | 1201 | |||
XM_005253965.4 | XP_005254022.1 | 10 | 1029 | |||
XM_006719653.3 | XP_006719716.1 | 12 | 1291 | |||
XM_011538894.2 | XP_011537196.1 | 9 | 922 | |||
XM_017020048.1 | XP_016875537.1 | 11 | 1213 | |||
XM_017020049.1 | XP_016875538.1 | 11 | 1201 | |||
XM_017020050.1 | XP_016875539.1 | 11 | 1313 | |||
XM_017020051.1 | XP_016875540.1 | 12 | 1169 | |||
XM_017020052.1 | XP_016875541.1 | 11 | 1215 | |||
XM_017020053.1 | XP_016875542.1 | 10 | 945 | |||
XM_017020054.1 | XP_016875543.1 | 9 | 820 | |||
GenBank mRNA | AJ586136.1 | - | 11 | 1109 | ||
AK027820.1 | - | 11 | 1183 | |||
AL834278.1 | - | 2 | 381 | |||
BC004868.2 | - | 11 | 1208 | 3833242 | ||
BC022094.2 | - | 9 | 760 | |||
CN367077.1 | - | 5 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
