siRNA Details
Gene Symbol: UBE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130466.3 | NP_569733.2 | 28 | 4193 | ||
NM_183415.2 | NP_904324.1 | 28 | 3848 | |||
XM_005253987.2 | XP_005254044.1 | 28 | 4073 | |||
XM_006719681.3 | XP_006719744.1 | 28 | 5052 | |||
XM_006719682.2 | XP_006719745.1 | 28 | 5049 | |||
XM_011538959.2 | XP_011537261.1 | 27 | 3955 | |||
XM_017020195.1 | XP_016875684.1 | 28 | 4616 | |||
GenBank mRNA | AF251046.2 | - | 28 | 4163 | ||
AK056015.1 | - | 1 | 461 | |||
AK127227.1 | - | 13 | 2980 | |||
BC141880.1 | - | 28 | 4071 | 40124513 | ||
BC144684.1 | - | 28 | 4169 | |||
BC150637.1 | - | 28 | 3824 | 9057007 | ||
BX537403.1 | - | 28 | 3838 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
