siRNA Details
Gene Symbol: UBE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130466.3 | NP_569733.2 | 13, 14 | 1898 | ||
NM_183415.2 | NP_904324.1 | 13, 14 | 1553 | |||
XM_005253987.2 | XP_005254044.1 | 13, 14 | 1778 | |||
XM_006719681.3 | XP_006719744.1 | 13, 14 | 2111 | |||
XM_006719682.2 | XP_006719745.1 | 13, 14 | 2111 | |||
XM_011538959.2 | XP_011537261.1 | 12, 13 | 1660 | |||
XM_011538961.1 | XP_011537263.1 | 13, 14 | 2111 | |||
XM_017020195.1 | XP_016875684.1 | 13, 14 | 2321 | |||
GenBank mRNA | AF130072.1 | - | 11, 12 | 1290 | ||
AF251046.2 | - | 13, 14 | 1868 | |||
AK307385.1 | - | 12, 13 | 1432 | |||
AL096740.1 | - | 13, 14 | 1867 | |||
BC032301.1 | - | 13, 14 | 1531 | |||
BC141880.1 | - | 13, 14 | 1776 | 40124513 | ||
BC144684.1 | - | 13, 14 | 1874 | |||
BC150637.1 | - | 13, 14 | 1529 | 9057007 | ||
BX537403.1 | - | 13, 14 | 1543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
