siRNA Details
Gene Symbol: SETD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171990.2 | NP_001165461.1 | 5 | 1104 | ||
NM_001323018.1 | NP_001309947.1 | 5 | 953 | |||
NM_001323022.1 | NP_001309951.1 | 5 | 681 | |||
NM_153706.3 | NP_714917.2 | 5 | 1104 | |||
NR_136555.1 | 4 | 736 | ||||
NR_136556.1 | 4 | 736 | ||||
NR_136557.1 | 4 | 585 | ||||
NR_136558.1 | 4 | 736 | ||||
XM_005248426.4 | XP_005248483.1 | 4 | 404 | |||
XM_011543139.2 | XP_011541441.1 | 5 | 1118 | |||
XM_011543140.2 | XP_011541442.1 | 5 | 732 | |||
GenBank mRNA | AK309152.1 | - | 5 | 1012 | ||
BC036528.1 | - | 5 | 1091 | 4827793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
