siRNA Details
Gene Symbol: SETD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171990.2 | NP_001165461.1 | 3 | 877 | ||
NM_001323018.1 | NP_001309947.1 | 3 | 726 | |||
NM_001323019.1 | NP_001309948.1 | 3 | 877 | |||
NM_001323020.1 | NP_001309949.1 | 3 | 726 | |||
NM_001323022.1 | NP_001309951.1 | 3 | 454 | |||
NM_153706.3 | NP_714917.2 | 3 | 877 | |||
NR_136555.1 | 2 | 509 | ||||
NR_136556.1 | 2 | 509 | ||||
NR_136557.1 | 2 | 358 | ||||
NR_136558.1 | 2 | 509 | ||||
XM_005248426.4 | XP_005248483.1 | 2 | 177 | |||
XM_011543139.2 | XP_011541441.1 | 3 | 891 | |||
XM_011543140.2 | XP_011541442.1 | 3 | 505 | |||
GenBank mRNA | AK309152.1 | - | 3 | 785 | ||
BC036528.1 | - | 3 | 864 | 4827793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
