siRNA Details
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 9 | 1169 | ||
NM_182931.2 | NP_891847.1 | 10 | 1317 | |||
XM_005250493.1 | XP_005250550.1 | 9 | 1236 | |||
XM_011516400.1 | XP_011514702.1 | 11 | 1443 | |||
XM_017012435.1 | XP_016867924.1 | 10 | 1317 | |||
GenBank mRNA | AF067804.1 | - | 6 | 636 | ||
AF519459.1 | - | 9 | 984 | |||
AK000940.1 | - | 9 | 984 | |||
AK024088.1 | - | 2342 | ||||
AY147037.1 | - | 10 | 1317 | |||
AY157990.1 | - | 8 | 783 | |||
BC001296.2 | - | 10 | 1279 | |||
BC016382.1 | - | 6 | 482 | |||
BC024658.1 | - | 6 | 612 | |||
BC062583.1 | - | 10 | 1289 | |||
BC142987.1 | - | 9 | 1199 | |||
JQ809698.1 | - | 8 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
