siRNA Details
Gene Symbol: UBE2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058167.2 | NP_477515.2 | 7 | 1077 | ||
NM_194315.1 | NP_919296.1 | 8 | 1125 | |||
NM_194457.1 | NP_919439.1 | 6 | 946 | |||
NM_194458.1 | NP_919440.1 | 7 | 1206 | |||
XM_005244718.3 | XP_005244775.1 | 7 | 1355 | |||
XM_005244719.4 | XP_005244776.1 | 7 | 1243 | |||
XM_006710333.3 | XP_006710396.1 | 7 | 1522 | |||
XM_011540613.2 | XP_011538915.1 | 6 | 950 | |||
XM_011540614.2 | XP_011538916.1 | 7 | 1308 | |||
XM_011540616.2 | XP_011538918.1 | 8 | 1317 | |||
XM_011540617.2 | XP_011538919.1 | 7 | 1084 | |||
XM_017000238.1 | XP_016855727.1 | 8 | 1188 | |||
XM_017000239.1 | XP_016855728.1 | 6 | 1406 | |||
GenBank mRNA | AF296658.1 | - | 7 | 1026 | ||
AK056065.1 | - | 7 | 987 | |||
AK075017.1 | - | 7 | 1056 | |||
AK098803.1 | - | 2 | 443 | |||
AK291935.1 | - | 7 | 1056 | |||
AW411508.1 | - | 3 | 605 | |||
BC094777.1 | - | 7 | 1076 | 30340049 | ||
BC104890.1 | - | 6 | 901 | 8143893 | ||
BC113430.1 | - | 6 | 901 | 8322482 | ||
BC143657.1 | - | 6 | 901 | 9052171 | ||
BQ881730.1 | - | 5 | 766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
