siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270453.1 | NP_001257382.1 | 20 | 2278 | ||
NM_001270454.1 | NP_001257383.1 | 23 | 2588 | |||
NM_007014.4 | NP_008945.2 | 24 | 2641 | |||
NM_199424.2 | NP_955456.1 | 13 | 1733 | |||
XM_011522823.2 | XP_011521125.1 | 25 | 2671 | |||
XM_011522825.1 | XP_011521127.1 | 23 | 2552 | |||
XM_011522826.2 | XP_011521128.1 | 20 | 2442 | |||
XM_017022879.1 | XP_016878368.1 | 25 | 2745 | |||
XM_017022880.1 | XP_016878369.1 | 25 | 2677 | |||
XM_017022881.1 | XP_016878370.1 | 25 | 2721 | |||
GenBank mRNA | AK126332.1 | - | 13 | 1717 | ||
AK295070.1 | - | 16 | 1633 | |||
AK300266.1 | - | 20 | 2278 | |||
AK312792.1 | - | 23 | 2568 | |||
AK316097.1 | - | 14 | 1587 | |||
BC013645.1 | - | 22 | 2475 | 3453212 | ||
BC064531.1 | - | 23 | 2527 | 5588092 | ||
JN712744.1 | - | 23 | 2558 | |||
U96114.1 | - | 24 | 2573 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
