siRNA Details
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281742.1 | NP_001268671.1 | 3 | 297 | ||
NM_007019.3 | NP_008950.1 | 3 | 297 | |||
NM_181799.2 | NP_861515.1 | 3 | 297 | |||
NM_181801.3 | NP_861517.1 | 3 | 334 | |||
GenBank mRNA | AK311949.1 | - | 3 | 264 | ||
BC007656.2 | - | 3 | 220 | 3345575 | ||
BC016292.1 | - | 3 | 226 | 3910286 | ||
BC028161.1 | - | 2 | 836 | |||
BC050736.1 | - | 3 | 247 | 6503915 | ||
BF974975.1 | - | 353 | ||||
BI858659.1 | - | 3 | 334 | |||
BM556795.1 | - | 3 | 215 | |||
BM747513.1 | - | 3 | 252 | |||
BP227692.1 | - | 3 | 324 | |||
BT007300.1 | - | 3 | 177 | |||
BT020165.1 | - | 3 | 177 | |||
BT020166.1 | - | 3 | 177 | |||
BU169315.1 | - | 3 | 238 | |||
CN354813.1 | - | 3 | 198 | |||
CR456988.1 | - | 3 | 177 | |||
DB062573.1 | - | 3 | 298 | |||
U73379.1 | - | 3 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
