siRNA Details
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 2 | 409 | ||
XM_011535887.1 | XP_011534189.1 | 2 | 396 | |||
XM_011535888.2 | XP_011534190.1 | 2 | 426 | |||
GenBank mRNA | AF151834.1 | - | 2 | 216 | ||
AF161502.1 | - | 2 | 104 | |||
AJ245898.1 | - | 2 | 140 | |||
AJ420578.1 | - | 1 | 58 | |||
AK223464.1 | - | 3 | 148 | |||
AK290574.1 | - | 2 | 251 | |||
AK300629.1 | - | 3 | 154 | |||
BC013973.1 | - | 2 | 371 | 4137664 | ||
BF058850.1 | - | 3 | 328 | |||
U93243.1 | - | 2 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
