siRNA Details
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 5 | 715 | ||
XM_011535887.1 | XP_011534189.1 | 5 | 702 | |||
XM_011535888.2 | XP_011534190.1 | 5 | 732 | |||
GenBank mRNA | AF151039.1 | - | 2 | 109 | ||
AF151834.1 | - | 5 | 522 | |||
AF161502.1 | - | 5 | 410 | |||
AJ245898.1 | - | 5 | 446 | |||
AJ420578.1 | - | 4 | 364 | |||
AK223464.1 | - | 6 | 454 | |||
AK290574.1 | - | 5 | 557 | |||
AK300629.1 | - | 6 | 460 | |||
BC013973.1 | - | 5 | 677 | 4137664 | ||
U93243.1 | - | 5 | 411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
