siRNA Details
Gene Symbol: UBE2J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016021.2 | NP_057105.2 | 8 | 1914 | ||
XM_011535887.1 | XP_011534189.1 | 7 | 1781 | |||
XM_011535888.2 | XP_011534190.1 | 8 | 1961 | |||
GenBank mRNA | AF151039.1 | - | 5 | 1308 | ||
AF151834.1 | - | 8 | 1721 | |||
AJ420578.1 | - | 7 | 1564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
