siRNA Details
Gene Symbol: WHSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042424.2 | NP_001035889.1 | 3 | 927 | ||
NM_007331.1 | NP_015627.1 | 5 | 1245 | |||
NM_133330.2 | NP_579877.1 | 5 | 1245 | |||
NM_133331.2 | NP_579878.1 | 4 | 1065 | |||
NM_133334.2 | NP_579889.1 | 2 | 780 | |||
NM_133335.3 | NP_579890.1 | 3 | 890 | |||
XM_005248001.3 | XP_005248058.1 | 3 | 870 | |||
XM_005248002.2 | XP_005248059.1 | 3 | 966 | |||
XM_005248005.2 | XP_005248062.1 | 3 | 966 | |||
XM_006713914.3 | XP_006713977.1 | 4 | 1135 | |||
XM_011513557.2 | XP_011511859.1 | 3 | 806 | |||
GenBank mRNA | AF071593.1 | - | 2 | 780 | ||
AF071594.1 | - | 2 | 780 | |||
AF083386.1 | - | 5 | 1245 | |||
AF083387.1 | - | 4 | 1065 | |||
AF083388.1 | - | 3 | 889 | |||
AF083389.1 | - | 5 | 1245 | |||
AF083390.1 | - | 5 | 1245 | |||
AF083391.1 | - | 5 | 1245 | |||
AJ007042.1 | - | 3 | 1513 | |||
AK289697.1 | - | 4 | 912 | |||
AY694128.1 | - | 2 | 762 | |||
BC020545.1 | - | 3 | 862 | |||
BC052254.1 | - | 3 | 869 | |||
BC141815.1 | - | 3 | 960 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
