siRNA Details
Gene Symbol: UBE2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281741.1 | NP_001268670.1 | 5 | 623 | ||
NM_001281742.1 | NP_001268671.1 | 5 | 559 | |||
NM_007019.3 | NP_008950.1 | 6 | 764 | |||
NM_181799.2 | NP_861515.1 | 6 | 710 | |||
NM_181800.2 | NP_861516.1 | 5 | 677 | |||
NM_181801.3 | NP_861517.1 | 6 | 801 | |||
NR_104036.1 | 4 | 472 | ||||
NR_104037.1 | 5 | 567 | ||||
GenBank mRNA | BC007656.2 | - | 6 | 687 | 3345575 | |
BC016292.1 | - | 6 | 693 | 3910286 | ||
BC028161.1 | - | 5 | 1303 | |||
BC050736.1 | - | 6 | 714 | 6503915 | ||
BM556795.1 | - | 6 | 628 | |||
BU171488.1 | - | 5 | 583 | |||
BU844974.1 | - | 5 | 579 | |||
CN354813.1 | - | 5 | 460 | |||
CN354815.1 | - | 5 | 502 | |||
U73379.1 | - | 6 | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
