siRNA Details
Gene Symbol: BIRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012270.1 | NP_001012270.1 | 3 | 1351 | ||
NM_001012271.1 | NP_001012271.1 | 5 | 1538 | |||
NM_001168.2 | NP_001159.2 | 4 | 1469 | |||
GenBank mRNA | AF077350.1 | - | 4 | 1372 | ||
AK223428.1 | - | 4 | 1415 | |||
AL519719.3 | - | 1 | 44 | |||
BC000784.2 | - | 3 | 1270 | |||
BC008718.2 | - | 4 | 1381 | 2961114 | ||
BC034148.1 | - | 4 | 1386 | 4656567 | ||
BC065497.1 | - | 4 | 1361 | 5394399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
