siRNA Details
Gene Symbol: PRDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198.3 | NP_001189.2 | 4 | 821 | ||
NM_182907.2 | NP_878911.1 | 2 | 409 | |||
XM_006715550.3 | XP_006715613.1 | 4 | 1799 | |||
XM_011536062.2 | XP_011534364.1 | 4 | 1795 | |||
XM_011536063.2 | XP_011534365.1 | 4 | 873 | |||
XM_011536064.2 | XP_011534366.1 | 2 | 1974 | |||
XM_017011187.1 | XP_016866676.1 | 4 | 608 | |||
GenBank mRNA | AF084199.1 | - | 4 | 702 | ||
AK289556.1 | - | 4 | 765 | |||
AK301341.1 | - | 2 | 409 | |||
AL832963.1 | - | 4 | 755 | |||
AY198414.1 | - | 4 | 821 | |||
AY198415.1 | - | 2 | 331 | |||
BC103832.1 | - | 2 | 322 | 40001920 | ||
BC103833.1 | - | 2 | 322 | 40001921 | ||
BC103834.1 | - | 2 | 322 | 40001922 | ||
BC103835.1 | - | 2 | 322 | 40001923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
