siRNA Details
Gene Symbol: PRDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198.3 | NP_001189.2 | 7 | 4481 | ||
NM_182907.2 | NP_878911.1 | 5 | 4069 | |||
XM_006715550.3 | XP_006715613.1 | 6 | 5330 | |||
XM_011536062.2 | XP_011534364.1 | 7 | 5455 | |||
XM_011536063.2 | XP_011534365.1 | 7 | 4533 | |||
XM_011536064.2 | XP_011534366.1 | 5 | 5634 | |||
XM_017011187.1 | XP_016866676.1 | 7 | 4268 | |||
GenBank mRNA | AK129768.1 | - | 1 | 1257 | ||
AY198414.1 | - | 7 | 4481 | |||
AY198415.1 | - | 5 | 3991 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
