siRNA Details
Gene Symbol: CDK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145306.1 | NP_001138778.1 | 8 | 4811 | ||
NM_001259.6 | NP_001250.1 | 8 | 4706 | |||
XM_006715835.2 | XP_006715898.1 | 8 | 4931 | |||
GenBank mRNA | AK172791.1 | - | 1 | 2102 | ||
BM905472.1 | - | 1 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
