siRNA Details
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 727 | ||
NM_003336.3 | NP_003327.2 | 6 | 734 | |||
NM_181762.2 | NP_861427.1 | 5 | 644 | |||
GenBank mRNA | AK223045.1 | - | 6 | 636 | ||
AK223064.1 | - | 6 | 633 | |||
AK301675.1 | - | 5 | 645 | |||
BC010175.2 | - | 6 | 627 | 3051041 | ||
BC042021.1 | - | 3 | 1511 | |||
BI093289.1 | - | 5 | 434 | |||
M74524.1 | - | 6 | 609 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
