siRNA Details
Gene Symbol: UBE2G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003342.4 | NP_003333.1 | 6 | 1118 | ||
GenBank mRNA | BC002775.2 | - | 6 | 1017 | 3619626 | |
BC026288.1 | - | 6 | 926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
