siRNA Details
Gene Symbol: UBE2G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202489.1 | NP_001189418.1 | 4 | 2715 | ||
NM_003343.5 | NP_003334.2 | 6 | 2797 | |||
NM_182688.2 | NP_872630.1 | 7 | 2912 | |||
XM_017028469.1 | XP_016883958.1 | 6 | 2756 | |||
GenBank mRNA | AF032456.1 | - | 6 | 2765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
