siRNA Details
Gene Symbol: UBE2V2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003350.2 | NP_003341.1 | 4 | 1032 | ||
XM_005251300.2 | XP_005251357.1 | 4 | 1091 | |||
XM_011517583.2 | XP_011515885.1 | 4 | 1110 | |||
XM_017013808.1 | XP_016869297.1 | 4 | 1017 | |||
GenBank mRNA | AF049140.1 | - | 4 | 1032 | ||
AK094617.1 | - | 3 | 2281 | |||
BC007051.1 | - | 4 | 1037 | 3683051 | ||
BC016332.1 | - | 4 | 1057 | 4109228 | ||
BC016710.1 | - | 4 | 1041 | 4108684 | ||
BC028673.1 | - | 4 | 1034 | 4104992 | ||
BC062418.1 | - | 4 | 1031 | 6503261 | ||
U62136.2 | - | 4 | 1033 | |||
X98091.1 | - | 4 | 1057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
