siRNA Details
Gene Symbol: KMT2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003482.3 | NP_003473.3 | 39 | 12073 | ||
XM_005269162.4 | XP_005269219.1 | 40 | 13352 | |||
XM_006719614.3 | XP_006719677.1 | 40 | 13361 | |||
XM_006719616.3 | XP_006719679.1 | 40 | 13349 | |||
XM_011538770.2 | XP_011537072.1 | 40 | 13364 | |||
XM_011538771.2 | XP_011537073.1 | 40 | 13361 | |||
XM_011538772.2 | XP_011537074.1 | 40 | 13355 | |||
XM_011538773.2 | XP_011537075.1 | 40 | 13352 | |||
XM_011538774.2 | XP_011537076.1 | 40 | 13344 | |||
XM_011538776.2 | XP_011537078.1 | 40 | 13271 | |||
GenBank mRNA | AB209494.1 | - | 6 | 3575 | ||
AF010403.1 | - | 39 | 11248 | |||
AF010404.1 | - | 30 | 10333 | |||
X85325.1 | - | 1 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
