siRNA Details
Gene Symbol: KMT2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003482.3 | NP_003473.3 | 41, 42 | 13835 | ||
XM_005269162.4 | XP_005269219.1 | 42, 43 | 15114 | |||
XM_006719614.3 | XP_006719677.1 | 42, 43 | 15123 | |||
XM_006719616.3 | XP_006719679.1 | 42, 43 | 15111 | |||
XM_011538770.2 | XP_011537072.1 | 42, 43 | 15126 | |||
XM_011538771.2 | XP_011537073.1 | 42, 43 | 15123 | |||
XM_011538772.2 | XP_011537074.1 | 42, 43 | 15117 | |||
XM_011538773.2 | XP_011537075.1 | 42, 43 | 15114 | |||
XM_011538774.2 | XP_011537076.1 | 42, 43 | 15106 | |||
XM_011538776.2 | XP_011537078.1 | 42, 43 | 15033 | |||
GenBank mRNA | AB209494.1 | - | 8, 9 | 5337 | ||
AF010403.1 | - | 41, 42 | 13010 | |||
AF010404.1 | - | 32, 33 | 12095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
