siRNA Details
Gene Symbol: RUVBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319084.1 | NP_001306013.1 | 10 | 1350 | ||
NM_003707.2 | NP_003698.1 | 11 | 1383 | |||
XM_005247841.3 | XP_005247898.1 | 11 | 1304 | |||
GenBank mRNA | AB012122.1 | - | 11 | 1332 | ||
AB451224.1 | - | 11 | 1279 | |||
AB451347.1 | - | 11 | 1279 | |||
AF070735.1 | - | 11 | 1355 | |||
AF099084.1 | - | 11 | 1343 | |||
AJ010058.1 | - | 11 | 1355 | |||
AK222563.1 | - | 11 | 1364 | |||
AK222575.1 | - | 11 | 1355 | |||
AK312290.1 | - | 11 | 1379 | |||
BC002993.1 | - | 11 | 1350 | 2823568 | ||
BC012886.1 | - | 11 | 1312 | 3876400 | ||
BT007057.1 | - | 11 | 1279 | |||
DQ469310.1 | - | 10 | 1187 | |||
Y18418.1 | - | 11 | 1375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
