siRNA Details
Gene Symbol: RUVBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319084.1 | NP_001306013.1 | 4 | 616 | ||
NM_001319086.1 | NP_001306015.1 | 4 | 1024 | |||
NM_003707.2 | NP_003698.1 | 4 | 557 | |||
XM_005247841.3 | XP_005247898.1 | 4 | 478 | |||
XM_011513248.1 | XP_011511550.1 | 4 | 604 | |||
XM_011513249.2 | XP_011511551.1 | 4 | 611 | |||
XM_017007356.1 | XP_016862845.1 | 4 | 611 | |||
XM_017007357.1 | XP_016862846.1 | 4 | 481 | |||
GenBank mRNA | AB012122.1 | - | 4 | 506 | ||
AB451224.1 | - | 4 | 453 | |||
AB451347.1 | - | 4 | 453 | |||
AF070735.1 | - | 4 | 529 | |||
AF099084.1 | - | 4 | 517 | |||
AJ010058.1 | - | 4 | 529 | |||
AK092163.1 | - | 4 | 1024 | |||
AK222563.1 | - | 4 | 538 | |||
AK222575.1 | - | 4 | 529 | |||
AK312290.1 | - | 4 | 553 | |||
BC002993.1 | - | 4 | 524 | 2823568 | ||
BC012886.1 | - | 4 | 486 | 3876400 | ||
BT007057.1 | - | 4 | 453 | |||
BX447686.2 | - | 4 | 634 | |||
DA530328.1 | - | 4 | 557 | |||
DQ469310.1 | - | 4 | 453 | |||
Y18418.1 | - | 4 | 549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
