siRNA Details
Gene Symbol: HDAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003883.3 | NP_003874.2 | 13 | 1105 | ||
XM_011537697.2 | XP_011535999.1 | 12 | 957 | |||
GenBank mRNA | AF005482.1 | - | 11 | 1121 | ||
AF039703.1 | - | 13 | 1061 | |||
BC000614.1 | - | 13 | 1094 | 3345755 | ||
U66914.1 | - | 13 | 1050 | |||
U75696.1 | - | 12 | 1088 | |||
U75697.1 | - | 13 | 1094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
