siRNA Details
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257191.2 | NP_001244120.1 | 14 | 3000 | ||
NM_004397.5 | NP_004388.2 | 14 | 3072 | |||
XM_005271417.3 | XP_005271474.1 | 14 | 3017 | |||
XM_011542644.1 | XP_011540946.1 | 13 | 2943 | |||
XM_011542645.1 | XP_011540947.1 | 13 | 2925 | |||
XM_017017251.1 | XP_016872740.1 | 13 | 2888 | |||
GenBank mRNA | BC039826.1 | - | 7 | 1856 | ||
D17532.1 | - | 14 | 2943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
