siRNA Details
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 25 | 3885 | ||
XM_005268276.4 | XP_005268333.1 | 24 | 4015 | |||
XM_006720726.3 | XP_006720789.1 | 25 | 3891 | |||
XM_006720727.3 | XP_006720790.1 | 23 | 3648 | |||
XM_011522131.2 | XP_011520433.1 | 23 | 3672 | |||
XM_011522133.2 | XP_011520435.1 | 6 | 651 | |||
XM_017022695.1 | XP_016878184.1 | 24 | 3832 | |||
XM_017022696.1 | XP_016878185.1 | 24 | 3775 | |||
GenBank mRNA | AF071172.3 | - | 25 | 3837 |
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 6 | 712 | |||
GenBank mRNA | AB002391.2 | - | 4 | 457 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
6 | 716 | ||
AF041081.1 | - | LIT2191 HERC2P3 |
4 | 392 | ||
AK307870.1 | - | 1 | 95 | |||
AK309255.1 | - | 1 | 97 |
Gene Symbol: HERC2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036432.1 | 5 | 551 | |||
GenBank mRNA | AK294470.1 | - | 1 | 123 | ||
AK294795.1 | - | 1 | 97 | |||
BC033888.1 | - | 5 | 556 | |||
BC094757.1 | - | 5 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
