siRNA Details
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 27, 28 | 4329 | ||
XM_005268276.4 | XP_005268333.1 | 26, 27 | 4459 | |||
XM_006720726.3 | XP_006720789.1 | 27, 28 | 4335 | |||
XM_006720727.3 | XP_006720790.1 | 25, 26 | 4092 | |||
XM_011522131.2 | XP_011520433.1 | 25, 26 | 4116 | |||
XM_011522133.2 | XP_011520435.1 | 8, 9 | 1095 | |||
XM_017022695.1 | XP_016878184.1 | 26, 27 | 4276 | |||
XM_017022696.1 | XP_016878185.1 | 26, 27 | 4219 | |||
GenBank mRNA | AF071172.3 | - | 27, 28 | 4281 |
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 8, 9 | 1156 | |||
GenBank mRNA | AB002391.2 | - | 6, 7 | 901 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
8, 9 | 1160 | ||
AF041081.1 | - | LIT2191 HERC2P3 |
6, 7 | 836 | ||
AK307870.1 | - | 3, 4 | 538 |
Gene Symbol: HERC2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036432.1 | 7, 8 | 995 | |||
GenBank mRNA | AK294470.1 | - | 2, 3 | 416 | ||
AK294795.1 | - | 3, 4 | 541 | |||
BC033888.1 | - | 7, 8 | 1000 | |||
BC094757.1 | - | 7, 8 | 995 |
Gene Symbol: HERC2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM464322.1 | - | 1, 2 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
