siRNA Details
Gene Symbol: HDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004964.2 | NP_004955.2 | 14 | 1786 | ||
XM_011541309.2 | XP_011539611.1 | 14 | 2171 | |||
GenBank mRNA | AK225555.1 | - | 9 | 1038 | ||
AK299241.1 | - | 14 | 1720 | |||
BC000301.2 | - | 14 | 1757 | 2820260 | ||
BX648055.1 | - | 7 | 2089 | |||
D50405.1 | - | 14 | 1786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
