siRNA Details
Gene Symbol: USP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272075.1 | NP_001259004.1 | 13 | 2326 | ||
NM_005153.2 | NP_005144.2 | 12 | 2205 | |||
NR_073577.1 | 10 | 1103 | ||||
NR_073578.1 | 9 | 1942 | ||||
XM_006721332.1 | XP_006721395.1 | 13 | 2212 | |||
XM_011523440.1 | XP_011521742.1 | 14 | 2333 | |||
XM_011523441.1 | XP_011521743.1 | 15 | 2657 | |||
XM_011523443.1 | XP_011521745.1 | 14 | 2404 | |||
XM_017023863.1 | XP_016879352.1 | 13 | 2351 | |||
XM_017023864.1 | XP_016879353.1 | 16 | 2627 | |||
XM_017023865.1 | XP_016879354.1 | 15 | 2496 | |||
XM_017023866.1 | XP_016879355.1 | 13 | 3028 | |||
XM_017023867.1 | XP_016879356.1 | 15 | 2619 | |||
XM_017023868.1 | XP_016879357.1 | 14 | 2453 | |||
XM_017023869.1 | XP_016879358.1 | 14 | 2490 | |||
GenBank mRNA | AK293304.1 | - | 10 | 1049 | ||
AK299618.1 | - | 13 | 2271 | |||
AK315570.1 | - | 12 | 2159 | |||
AL162049.1 | - | 12 | 2144 | |||
BC000263.1 | - | 12 | 2176 | 3501606 | ||
BC064516.1 | - | 2 | 251 | |||
BX537402.1 | - | 12 | 2143 | |||
CR749515.1 | - | 9 | 1897 | |||
D80012.1 | - | 12 | 2109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
