siRNA Details
Gene Symbol: MCM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278595.1 | NP_001265524.1 | 5 | 1352 | ||
NM_005916.4 | NP_005907.3 | 6 | 1277 | |||
NM_182776.2 | NP_877577.1 | 5 | 1243 | |||
XM_005250348.3 | XP_005250405.1 | 6 | 931 | |||
XM_017012217.1 | XP_016867706.1 | 6 | 808 | |||
GenBank mRNA | AK055379.1 | - | 5 | 1220 | ||
AK096959.1 | - | 7 | 641 | |||
AK226175.1 | - | 5 | 782 | |||
AK293172.1 | - | 6 | 679 | |||
AK314575.1 | - | 6 | 744 | |||
BC009398.2 | - | 6 | 695 | 4134871 | ||
BC013375.2 | - | 6 | 713 | 3946342 | ||
D55716.1 | - | 6 | 747 | |||
D86748.1 | - | 6 | 717 | |||
KU178172.1 | - | 6 | 631 | |||
KU178173.1 | - | 5 | 706 | |||
X74796.1 | - | 3 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
